CH4017936922 ChemBridge Corporation

1-(2-fluorobenzyl)-2-methyl-1H-indole-3-carbaldehyde

  • MDL number MFCD02629521
  • Cas Number : 347323-87-9
  • Molecular Weight : 267.2976

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6554916

Safety Information

MSDS https://www.hit2lead.com/sds/6554916_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart