CH4017936981 ChemBridge Corporation

2-(2-chloro-6-ethoxy-4-formylphenoxy)propanoic acid

  • MDL number MFCD04083025
  • Cas Number : 590395-60-1
  • Molecular Weight : 272.6816

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6729959

Safety Information

MSDS https://www.hit2lead.com/sds/6729959_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart