CH4017936984 ChemBridge Corporation

2-(2-ethoxy-4-formylphenoxy)propanoic acid

  • MDL number MFCD04083022
  • Cas Number : 51264-80-3
  • Molecular Weight : 238.2366

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6758323

Safety Information

MSDS https://www.hit2lead.com/sds/6758323_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart