CH4017936986 ChemBridge Corporation

(4-bromo-2-formyl-6-methoxyphenoxy)acetic acid

  • MDL number MFCD04083027
  • Cas Number : 20037-38-1
  • Molecular Weight : 289.0795

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6763869

Safety Information

MSDS https://www.hit2lead.com/sds/6763869_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart