CH4017936996 ChemBridge Corporation

4-[(2-chloro-5-fluoropyrimidin-4-yl)oxy]-3-methoxybenzaldehyde

  • MDL number MFCD02630524
  • Cas Number : 525570-78-9
  • Molecular Weight : 282.6549

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6873131

Safety Information

MSDS https://www.hit2lead.com/sds/6873131_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart