CH4017937013 ChemBridge Corporation

1-(biphenyl-4-yloxy)acetone

  • MDL number MFCD02969026
  • Cas Number : 18859-38-6
  • Molecular Weight : 226.2705

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6976862

Safety Information

MSDS https://www.hit2lead.com/sds/6976862_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart