CH4017937025 ChemBridge Corporation

1-cyclopropyl-2,5-dimethyl-1H-pyrrole-3-carbaldehyde

  • MDL number MFCD04309657
  • Cas Number : 640264-45-5
  • Molecular Weight : 163.2163

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7061314

Safety Information

MSDS https://www.hit2lead.com/building-blocks/7061314
Availability details for
Quantity
  • Availability
  • Add to Cart