CH4019764078 ChemBridge Corporation

4-[(3,4-dichlorophenyl)sulfonyl]butan-2-one

  • MDL number MFCD00635452
  • Cas Number : 336195-38-1
  • Molecular Weight : 281.1556

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7089200

Safety Information

MSDS https://www.hit2lead.com/sds/7089200_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart