CH4025466667 ChemBridge Corporation

[3-(cyclopropylcarbonyl)-1H-indol-1-yl]acetic acid

  • MDL number MFCD05993133
  • Cas Number : 708295-12-9
  • Molecular Weight : 243.2579

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7862790

Safety Information

MSDS https://www.hit2lead.com/sds/7862790_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart