CH4025466906 ChemBridge Corporation

isopropyl (3-formyl-1H-indol-1-yl)acetate

  • MDL number MFCD05815946
  • Cas Number : 708991-26-8
  • Molecular Weight : 245.2738

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7865272

Safety Information

MSDS https://www.hit2lead.com/sds/7865272_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart