CH4025470856 ChemBridge Corporation

[4-amino-2-(methylamino)-1,3-thiazol-5-yl](phenyl)methanone

  • MDL number MFCD04060946
  • Cas Number : 107401-74-1
  • Molecular Weight : 233.2895

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7904092

Safety Information

MSDS https://www.hit2lead.com/sds/7904092_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart