CH4026585900 ChemBridge Corporation

2-(4-chlorophenyl)-3-methylquinoline-4-carboxylic acid

  • MDL number MFCD01114916
  • Cas Number : 43071-47-2
  • Molecular Weight : 297.7357

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3000593

Safety Information

MSDS https://www.hit2lead.com/sds/3000593_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart