CH4026585928 ChemBridge Corporation

5-ethylthiophene-3-carboxylic acid

  • MDL number MFCD03419849
  • Cas Number : 19156-51-5
  • Molecular Weight : 156.2022

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3007503

Safety Information

MSDS https://www.hit2lead.com/building-blocks/3007503
Availability details for
Quantity
  • Availability
  • Add to Cart