CH4026585932 ChemBridge Corporation

5-isopropylthiophene-3-carboxylic acid

  • MDL number MFCD03419855
  • Cas Number : 123418-51-9
  • Molecular Weight : 170.2288

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3007509

Safety Information

MSDS https://www.hit2lead.com/sds/3007509_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart