CH4026585934 ChemBridge Corporation

6-(1,1-dimethylpropyl)-1-benzothiophene-3-carboxylic acid

  • MDL number MFCD03419874
  • Cas Number : 667436-12-6
  • Molecular Weight : 248.3406

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3007530

Safety Information

MSDS https://www.hit2lead.com/building-blocks/3007530
Availability details for
Quantity
  • Availability
  • Add to Cart