CH4026585935 ChemBridge Corporation

5-methyl-4-phenylthiophene-3-carboxylic acid

  • MDL number MFCD03419883
  • Cas Number : 557792-56-0
  • Molecular Weight : 218.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin RU
Product Link https://www.hit2lead.com/building-blocks/3007539

Safety Information

MSDS https://www.hit2lead.com/sds/3007539_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart