CH4026585947 ChemBridge Corporation

8-methyl-2-(5-methyl-2-thienyl)quinoline-4-carboxylic acid

  • MDL number MFCD03420022
  • Cas Number : 667412-63-7
  • Molecular Weight : 283.3449

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3007736

Safety Information

MSDS https://www.hit2lead.com/building-blocks/3007736
Availability details for
Quantity
  • Availability
  • Add to Cart