CH4026585954 ChemBridge Corporation

2-(2-methoxyphenyl)-8-methylquinoline-4-carboxylic acid

  • MDL number MFCD03420035
  • Cas Number : 884497-38-5
  • Molecular Weight : 293.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin RU
Product Link https://www.hit2lead.com/building-blocks/3007758

Safety Information

MSDS https://www.hit2lead.com/sds/3007758_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart