CH4026585956 ChemBridge Corporation

2-(4-methoxyphenyl)-8-methylquinoline-4-carboxylic acid

  • MDL number MFCD03420036
  • Cas Number : 107027-47-4
  • Molecular Weight : 293.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin RU
Product Link https://www.hit2lead.com/building-blocks/3007760

Safety Information

MSDS https://www.hit2lead.com/sds/3007760_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart