CH4026585984 ChemBridge Corporation

6-methyl-2-pyridin-2-ylquinoline-4-carboxylic acid

  • MDL number MFCD01736600
  • Cas Number : 5110-01-0
  • Molecular Weight : 264.2787

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3013512

Safety Information

MSDS https://www.hit2lead.com/building-blocks/3013512
Availability details for
Quantity
  • Availability
  • Add to Cart