CH4026585995 ChemBridge Corporation

(4-bromo-2-chlorophenoxy)acetic acid

  • MDL number MFCD03422201
  • Cas Number : 77228-67-2
  • Molecular Weight : 265.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin RU
Product Link https://www.hit2lead.com/building-blocks/3013794

Safety Information

MSDS https://www.hit2lead.com/sds/3013794_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart