CH4026586000 ChemBridge Corporation

(2-chloro-4-nitrophenoxy)acetic acid

  • MDL number MFCD03414519
  • Molecular Weight : 231.5899

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3013802

Safety Information

MSDS https://www.hit2lead.com/building-blocks/3013802
Availability details for
Quantity
  • Availability
  • Add to Cart