CH4026586007 ChemBridge Corporation

[4-(1-methyl-1-phenylethyl)phenoxy]acetic acid

  • MDL number MFCD00423397
  • Cas Number : 70757-61-8
  • Molecular Weight : 270.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin RU
Product Link https://www.hit2lead.com/building-blocks/3013822

Safety Information

MSDS https://www.hit2lead.com/sds/3013822_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart