CH4026586009 ChemBridge Corporation

(2-isopropyl-5-methylphenoxy)acetic acid

  • MDL number MFCD00021757
  • Cas Number : 5333-40-4
  • Molecular Weight : 208.2536

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3013824

Safety Information

MSDS https://www.hit2lead.com/building-blocks/3013824
Availability details for
Quantity
  • Availability
  • Add to Cart