CH4026586014 ChemBridge Corporation

2-[3-(trifluoromethyl)phenoxy]propanoic acid

  • MDL number MFCD03422213
  • Cas Number : 836-35-1
  • Molecular Weight : 234.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin RU
Product Link https://www.hit2lead.com/building-blocks/3013831

Safety Information

MSDS https://www.hit2lead.com/sds/3013831_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart