CH4026586019 ChemBridge Corporation

2-(2-methylphenoxy)propanoic acid

  • MDL number MFCD01122029
  • Cas Number : 7345-21-3
  • Molecular Weight : 180.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin RU
Product Link https://www.hit2lead.com/building-blocks/3013838

Safety Information

MSDS https://www.hit2lead.com/sds/3013838_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart