CH4026586061 ChemBridge Corporation

2-(2-isopropylphenoxy)-2-methylpropanoic acid

  • MDL number MFCD03422289
  • Cas Number : 669726-38-9
  • Molecular Weight : 222.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin RU
Product Link https://www.hit2lead.com/building-blocks/3013947

Safety Information

MSDS https://www.hit2lead.com/sds/3013947_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart