CH4026586072 ChemBridge Corporation

2-(3-bromophenoxy)propanoic acid

  • MDL number MFCD01910032
  • Cas Number : 41183-67-9
  • Molecular Weight : 245.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin RU
Product Link https://www.hit2lead.com/building-blocks/3013973

Safety Information

MSDS https://www.hit2lead.com/sds/3013973_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart