CH4026586080 ChemBridge Corporation

2-{[3-(trifluoromethyl)benzyl]oxy}benzaldehyde

  • MDL number MFCD03422400
  • Cas Number : 667437-45-8
  • Molecular Weight : 280.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin RU
Product Link https://www.hit2lead.com/building-blocks/3014100

Safety Information

MSDS https://www.hit2lead.com/sds/3014100_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart