CH4026586101 ChemBridge Corporation

1-(3-methoxybenzyl)-1H-indole-3-carbaldehyde

  • MDL number MFCD03422507
  • Cas Number : 261637-72-3
  • Molecular Weight : 265.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin RU
Product Link https://www.hit2lead.com/building-blocks/3014331

Safety Information

MSDS https://www.hit2lead.com/sds/3014331_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart