CH4026586109 ChemBridge Corporation

cyclopentanecarbohydrazide

  • MDL number MFCD03002174
  • Molecular Weight : 128.1723

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3014983

Safety Information

MSDS https://www.hit2lead.com/sds/3014983_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart