CH4026586114 ChemBridge Corporation

2-(3-methylphenoxy)acetohydrazide

  • MDL number MFCD00553726
  • Cas Number : 36304-38-8
  • Molecular Weight : 180.2038

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3015212

Safety Information

MSDS https://www.hit2lead.com/building-blocks/3015212
Availability details for
Quantity
  • Availability
  • Add to Cart