CH4026586129 ChemBridge Corporation

2-[4-(1-methyl-1-phenylethyl)phenoxy]propanohydrazide

  • MDL number MFCD03423136
  • Cas Number : 70757-69-6
  • Molecular Weight : 298.3795

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3015269

Safety Information

MSDS https://www.hit2lead.com/building-blocks/3015269
Availability details for
Quantity
  • Availability
  • Add to Cart