CH4026586316 ChemBridge Corporation

(5E)-5-(4-hydroxy-3-methoxybenzylidene)-2-mercapto-1,3-thiazol-4(5H)-one

  • MDL number MFCD03812608
  • Cas Number : 5447-37-0
  • Molecular Weight : 267.3241

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3018319

Safety Information

signalword Warning
MSDS https://www.hit2lead.com/building-blocks/3018319
hstatements H302, H315, H319, H335.
Availability details for
Quantity
  • Availability
  • Add to Cart