CH4026586417 ChemBridge Corporation

2'-methoxybiphenyl-3-carboxylic acid

  • MDL number MFCD03424597
  • Cas Number : 168618-47-1
  • Molecular Weight : 228.2433

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4400431

Safety Information

MSDS https://www.hit2lead.com/sds/4400431_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart