CH4026586424 ChemBridge Corporation

5-(3-methoxyphenyl)-2-furoic acid

  • MDL number MFCD05022489
  • Cas Number : 54022-96-7
  • Molecular Weight : 218.2054

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4401949

Safety Information

MSDS https://www.hit2lead.com/sds/4401949_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart