CH4026586433 ChemBridge Corporation

3-(isopropylamino)propan-1-ol

  • MDL number MFCD00483086
  • Cas Number : 33918-15-9
  • Molecular Weight : 117.1894

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5116284

Safety Information

MSDS https://www.hit2lead.com/sds/5116284_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart