CH4026586449 ChemBridge Corporation

N-benzyl-1-phenylbut-3-en-1-amine

  • MDL number MFCD00757007
  • Cas Number : 88381-98-0
  • Molecular Weight : 237.3395

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5321032

Safety Information

MSDS https://www.hit2lead.com/sds/5321032_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart