CH4026586491 ChemBridge Corporation

1-(5-methyl-3-nitro-1H-pyrazol-1-yl)acetone

  • MDL number MFCD01243523
  • Cas Number : 957511-87-4
  • Molecular Weight : 183.1647

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5904367

Safety Information

MSDS https://www.hit2lead.com/sds/5904367_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart