CH4026586494 ChemBridge Corporation

(4-chloro-3,5-dimethyl-1H-pyrazol-1-yl)acetic acid

  • MDL number MFCD01114908
  • Cas Number : 374913-86-7
  • Molecular Weight : 188.6116

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5909786

Safety Information

MSDS https://www.hit2lead.com/sds/5909786_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart