CH4026586507 ChemBridge Corporation

(2E,4E)-5-(4-chlorophenyl)penta-2,4-dienal

  • MDL number MFCD05863695
  • Cas Number : 49678-03-7
  • Molecular Weight : 192.6416

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6053417

Safety Information

MSDS https://www.hit2lead.com/sds/6053417_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart