CH4026586508 ChemBridge Corporation

4-ethyl-7-methyl-5-(2-oxopropoxy)-2H-chromen-2-one

  • MDL number MFCD02057050
  • Cas Number : 314742-55-7
  • Molecular Weight : 260.2851

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6136591

Safety Information

MSDS https://www.hit2lead.com/sds/6136591_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart