CH4026586509 ChemBridge Corporation

4-ethyl-7-(1-methyl-2-oxopropoxy)-2H-chromen-2-one

  • MDL number MFCD02080209
  • Cas Number : 314742-28-4
  • Molecular Weight : 260.2851

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6136642

Safety Information

MSDS https://www.hit2lead.com/sds/6136642_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart