CH4026586513 ChemBridge Corporation

7-methyl-5-(1-methyl-2-oxopropoxy)-4-propyl-2H-chromen-2-one

  • MDL number MFCD02057401
  • Cas Number : 314742-59-1
  • Molecular Weight : 288.3383

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6160499

Safety Information

MSDS https://www.hit2lead.com/sds/6160499_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart