CH4026586518 ChemBridge Corporation

[(6-chloro-4-methyl-2-oxo-2H-chromen-7-yl)oxy]acetic acid

  • MDL number MFCD01815621
  • Cas Number : 326102-48-1
  • Molecular Weight : 268.6499

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6238415

Safety Information

MSDS https://www.hit2lead.com/sds/6238415_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart