CH4026586521 ChemBridge Corporation

2-[(1,1-dioxidotetrahydro-3-thienyl)amino]butanoic acid

  • MDL number MFCD01085583
  • Cas Number : 51070-57-6
  • Molecular Weight : 221.274

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6385250

Safety Information

MSDS https://www.hit2lead.com/sds/6385250_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart