CH4026586536 ChemBridge Corporation

{2-chloro-4-[(E)-(2,5-dioxoimidazolidin-4-ylidene)methyl]phenoxy}acetic acid

  • MDL number MFCD03550165
  • Cas Number : 392696-01-4
  • Molecular Weight : 296.6633

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6478345

Safety Information

MSDS https://www.hit2lead.com/sds/6478345_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart