CH4026586546 ChemBridge Corporation

5-isopropyl-4-methoxy-2-methylbenzaldehyde

  • MDL number MFCD02629380
  • Cas Number : 105337-42-6
  • Molecular Weight : 192.2542

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6531894

Safety Information

MSDS https://www.hit2lead.com/sds/6531894_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart