CH4026586561 ChemBridge Corporation

3-[(1-phenyl-1H-tetrazol-5-yl)thio]propanoic acid

  • MDL number MFCD00772095
  • Cas Number : 1437-67-8
  • Molecular Weight : 250.277

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6700023

Safety Information

MSDS https://www.hit2lead.com/sds/6700023_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart