CH4026586565 ChemBridge Corporation

2-[4-(3-formyl-2,5-dimethyl-1H-pyrrol-1-yl)phenoxy]propanoic acid

  • MDL number MFCD03848136
  • Cas Number : 812642-70-9
  • Molecular Weight : 287.3105

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6725381

Safety Information

MSDS https://www.hit2lead.com/sds/6725381_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart